Anti-RNA Polymerase II, DNA Directed, Polypeptide L, 7.6kD Chicken Polyclonal Antibody
Catalog # USBIR2039-33
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:RNA Polymerase II, DNA Directed, Polypeptide L, 7.6kD
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Chicken
- Isotype:IgY
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Amino acid number:1 to 67
- Storage buffer:PBS, pH 7,2, 0,02% sodium azide.
- Molecular weight:7.6 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Recommended dilution: Western Blot: 1:1000-1:2000
Type: Primary
Antigen: RNA Polymerase II, DNA Directed, Polypeptide L, 7.6kD
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse, Rat