Anti-RNA Polymerase II, DNA-directed, Polypeptide G Mouse Monoclonal Antibody [clone: 8C292]
Catalog # USBIR2039-29G
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:RNA Polymerase II, DNA-directed, Polypeptide G
- Clonality:Monoclonal
- Clone:8C292
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,4, 1% BSA, 0,05% sodium azide.
- Storage temperature:Cool
- Concentration:~0,1 mg/ml
- Shipping temperature:Ice
- Tested applications:Dot Blot
- Purification:purified by Protein G chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Recommended dilution: Western Blot: 0.2-2?g/ml; Flow Cytometry: 0.5-2?g/sample
Type: Primary
Antigen: RNA Polymerase II, DNA-directed, Polypeptide G
Clonality: monoclonal
Clone: 8C292
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat