Order Entry
Czech Republic
ContactUsLinkComponent
Anti-Ribonucleoprotein Particles Mouse Monoclonal Antibody [clone: 58-15]
Anti-Ribonucleoprotein Particles Mouse Monoclonal Antibody [clone: 58-15]
Catalog # USBIR2028-75B
Supplier:  US Biological
Anti-Ribonucleoprotein Particles Mouse Monoclonal Antibody [clone: 58-15]
Catalog # USBIR2028-75B
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Ribonucleoprotein Particles
  • Clonality:
    Monoclonal
  • Clone:
    58-15
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgM
  • Form:
    liquid
  • Storage buffer:
    PBS, 0,05% sodium azide.
  • Storage temperature:
    Cool
  • Concentration:
    ~0,1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: Ribonucleoprotein Particles
Clonality: monoclonal
Clone: 58-15
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: