Anti-RND3 Mouse Monoclonal Antibody [clone: 1D2]
Catalog # USBIR1998-13E
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Rho family GTPase 3
- Antigen symbol:RND3
- Clonality:Monoclonal
- Clone:1D2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:1 to 245
- Storage buffer:PBS, pH 7,2.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Recommended dilution: Immunochemistry: 10 ?g/ml; Western Blot: 1:500-1:1000
Type: Primary
Antigen: RND3 (Rho family GTPase 3)
Clonality: monoclonal
Clone: 1D2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human