Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-QRFP Pyroglutamylated RFamide Peptide 43 Rabbit Polyclonal Antibody (Biotin)
Catalog # USBIQ0575-50B
Supplier:  US Biological
Anti-QRFP Pyroglutamylated RFamide Peptide 43 Rabbit Polyclonal Antibody (Biotin)
Catalog # USBIQ0575-50B
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    QRFP Pyroglutamylated RFamide Peptide 43
  • Clonality:
    Polyclonal
  • Conjugation:
    Biotin
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Form:
    liquid
  • Amino acid number:
    154 to 196
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

Type: Primary
Antigen: QRFP Pyroglutamylated RFamide Peptide 43
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human