Anti-Protein Phosphatase 2A, A Subunit Rabbit Polyclonal Antibody
Catalog # USBIP9107-34T
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Protein Phosphatase 2A, A Subunit
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Drosophila,Monkey,Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Storage buffer:10mM HEPES, pH 7,5, 150mM sodium chloride,0,1mg/ml BSA, 50% glycerol.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Recommended dilution: Western Blot: 1:1000; Immunofluorescence: 1:50; Flow Cytometry: 1:100
Type: Primary
Antigen: Protein Phosphatase 2A, A Subunit
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Drosophila, Human, Monkey, Mouse, Rat