Zadání objednávky
Czech Republic
ContactUsLinkComponent
Anti-Protein Phosphatase 2A, alpha, Catalytic Subunit Rat Monoclonal Antibody [clone: 8H123]
Anti-Protein Phosphatase 2A, alpha, Catalytic Subunit Rat Monoclonal Antibody [clone: 8H123]
Catalog # USBIP9107-32D
Supplier:  US Biological
Anti-Protein Phosphatase 2A, alpha, Catalytic Subunit Rat Monoclonal Antibody [clone: 8H123]
Catalog # USBIP9107-32D
Supplier:  US Biological

Specifications

  • Druh protilátky:
    Primární
  • Název antigenu:
    Protein Phosphatase 2A, alpha, Catalytic Subunit
  • Klonovalita:
    Monoclonal
  • Klon:
    8H123
  • Spojení:
    Unconjugated
  • Hostitel:
    Rat
  • Izotyp:
    IgG2b
  • Reaktivita:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Formulář:
    lyophilized powder
  • Skladovací pufr:
    PBS, 5% trehalose.
  • Skladovací teplota:
    Cool
  • Transportní teplota:
    Ice
  • Čištění:
    purified by Protein G chromatography
  • Rozměry protilátky:
    100 µg
  • Bal.:
    100 µG

Specifications

About this item

Recommended dilution: Western Blot: 1.0 ?g/ml is

Type: Primary
Antigen: Protein Phosphatase 2A, alpha, Catalytic Subunit
Clonality: monoclonal
Clone: 8H123
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Human, Mouse, Rat