Anti-Protein Phosphatase 2A, alpha, Catalytic Subunit Rat Monoclonal Antibody [clone: 8H124]
USBIP9107-32C
- Antibody type:Primary
- Antigen name:Protein Phosphatase 2A, alpha, Catalytic Subunit
- Clonality:Monoclonal
- Clone:8H124
- Conjugation:Unconjugated
- Host:Rat
- Isotype:IgG2b
- Reactivity:Human
- Form:lyophilized powder
- Amino acid number:209 to 309
- Storage buffer:PBS, 5% trehalose.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: Protein Phosphatase 2A, alpha, Catalytic Subunit
Clonality: monoclonal
Clone: 8H124
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Human