Anti-PROCR Rat Monoclonal Antibody [clone: LMR-42]
Catalog # USBIP9102-39L
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Protein C receptor, endothelial
- Antigen symbol:PROCR
- Clonality:Monoclonal
- Clone:LMR-42
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rat
- ImmunoChemistry:Yes
- Isotype:IgG2a
- Reactivity:Human
- Format:Supernatant
- Form:liquid
- Storage buffer:0,7%BSA, 0,1% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:1 ml
- Pk:1 mL
Specifications
About this item
Recommended dilution: Immunochemistry: 1:20
Type: Primary
Antigen: PROCR (protein C receptor, endothelial)
Clonality: monoclonal
Clone: LMR-42
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human