Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-Progesterone Mouse Monoclonal Antibody [clone: Pg.53]
  USBIP9006-02J
Anti-Progesterone Mouse Monoclonal Antibody [clone: Pg.53]
  USBIP9006-02J

 

  • Antibody type:
    Primary
  • Antigen symbol:
    Progesterone
  • Clonality:
    Monoclonal
  • Clone:
    Pg.53
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,4, 15mM sodium azide.
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Type: Primary
Antigen: Progesterone
Clonality: monoclonal
Clone: Pg.53
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human