Anti-PMAIP1 Rabbit Polyclonal Antibody
Catalog # USBIP4285-50D
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:phorbol-12-myristate-13-acetate-induced Protein 1
- Antigen symbol:PMAIP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Mouse
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, 0,02% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Recommended dilution: Western Blot: 0.5-1 ?g/ml; Immunochemistry: 5-10 ?g/ml
Type: Primary
Antigen: PMAIP1 (phorbol-12-myristate-13-acetate-induced protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse