Anti-Parainfluenza Virus Type 1 Mouse Monoclonal Antibody [clone: 10B2448]
Catalog # USBIP3107-05R
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Parainfluenza Virus Type 1
- Clonality:Monoclonal
- Clone:10B2448
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2b
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,09% sodium azide.
- Storage temperature:Cool
- Concentration:~0,1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: ELISA: 1:20-1:200; Immunofluorescence: 1:10-1:50
Type: Primary
Antigen: Parainfluenza Virus Type 1
Clonality: monoclonal
Clone: 10B2448
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: