Anti-Papilloma Virus, Human, Type 16/18, E7 Mouse Monoclonal Antibody [clone: 5E393]
Catalog # USBIP3105-19
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Papilloma Virus, Human, Type 16/18, E7
- Clonality:Monoclonal
- Clone:5E393
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,1% sodium azide.
- Storage temperature:Cool
- Concentration:~0,1 mg/ml
- Shipping temperature:Ice
- Purification:purified by protein A chromatography or sequential differential precipitation
- Size:1 ml
- Pk:1 mL
Specifications
About this item
Recommended dilution: ELISA: 1:20-1:200; Western Blot: 1:10-1:50
Type: Primary
Antigen: Papilloma Virus, Human, Type 16/18, E7
Clonality: Monoclonal
Clone: 5E393
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: