Anti-PACAP-38 Rabbit Polyclonal Antibody
Catalog # USBIP1775-03B
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:PACAP38
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Sheep,Human,Rat
- Format:Serum
- Form:lyophilized powder
- Antigen synonyms:PACAP-38
- Amino acid number:28 to 38
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Recommended dilution: Immunochemistry: 1:500
Type: Primary
Antigen: PACAP-38
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat, Sheep