Anti-Neuronostatin-13 Rabbit Polyclonal Antibody
USBIN2175-84A
- Antibody type:Primary
- Antigen name:Neuronostatin-13
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Pig,Human
- Form:lyophilized powder
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified
- Size:200 µg
- Pk:200 µG
Type: Primary
Antigen: Neuronostatin-13
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Pig