Anti-Nerve Growth Factor Receptor, p75NTR Mouse Monoclonal Antibody ATTO 488 [clone: 9G398]
USBIN2050-40C
- Antibody type:Primary
- Antigen name:Nerve Growth Factor Receptor, p75NTR
- Clonality:Monoclonal
- Clone:9G398
- Conjugation:ATTO 488
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a
- Reactivity:Rat
- Form:liquid
- Storage buffer:PBS, pH 7,2
- Storage temperature:Cool
- Shipping temperature:Ice
- Tested applications:Fluoresence ELISA
- Purification:purified by affinity chromatography
- Size:50 µg
- Pk:50 µG
Recommended dilution: Flow Cytometry: 20 ?g/ml; ELISA: 1:5000; Immunochemistry: 1-2?g/mI
Type: Primary
Antigen: Nerve Growth Factor Receptor, p75NTR
Clonality: monoclonal
Clone: 9G398
Conjugation: ATTO 488
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Rat