Order Entry
Czech Republic
ContactUsLinkComponent
Anti-Mucin 16 Mouse Monoclonal Antibody [clone: X306]
Anti-Mucin 16 Mouse Monoclonal Antibody [clone: X306]
Catalog # USBIM4701-95S
Supplier:  US Biological
Anti-Mucin 16 Mouse Monoclonal Antibody [clone: X306]
Catalog # USBIM4701-95S
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Mucin 16
  • Antigen symbol:
    MUC16
  • Clonality:
    Monoclonal
  • Clone:
    X306
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    TBS,0,09% sodium azide.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by ion exchange chromatography
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

Recommended dilution: Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: Mucin 16
Clonality: monoclonal
Clone: X306
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human