Anti-Mucin 16 Mouse Monoclonal Antibody [clone: 10B407]
Catalog # USBIM4701-95K
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Mucin 16
- Antigen symbol:MUC16
- Clonality:Monoclonal
- Clone:10B407
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:20mM Tris-phosphate, 100mM sodium chloride,0,09% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by ion exchange chromatography
- Size:200 µg
- Pk:200 µG
Specifications
About this item
Type: Primary
Antigen: Mucin 16
Clonality: monoclonal
Clone: 10B407
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human