Anti-Mucin 16 Mouse Monoclonal Antibody [clone: 10B406]
USBIM4701-95J
- Antibody type:Primary
- Antigen name:Mucin 16
- Antigen symbol:MUC16
- Clonality:Monoclonal
- Clone:10B406
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Form:liquid
- Storage buffer:20mM Tris-phosphate, 100mM sodium chloride.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:200 µg
- Pk:200 µG
Type: Primary
Antigen: Mucin 16
Clonality: monoclonal
Clone: 10B406
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human