Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-Mucin 16 Mouse Monoclonal Antibody [clone: 9H547]
  USBIM4701-95F
Anti-Mucin 16 Mouse Monoclonal Antibody [clone: 9H547]
  USBIM4701-95F

 

  • Antibody type:
    Primary
  • Antigen name:
    Mucin 16
  • Antigen symbol:
    MUC16
  • Clonality:
    Monoclonal
  • Clone:
    9H547
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2a
  • Reactivity:
    Human
  • Form:
    lyophilized powder
  • Storage buffer:
    0,01M PBS, pH 7,2.
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein G chromatography
  • Size:
    500 µg
  • Pk:
    500 µG

 

 

Type: Primary
Antigen: Mucin 16
Clonality: monoclonal
Clone: 9H547
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human