Anti-Mucin 16 Mouse Monoclonal Antibody [clone: 9H545]
Catalog # USBIM4701-95D
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Mucin 16
- Antigen symbol:MUC16
- Clonality:Monoclonal
- Clone:9H545
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Form:lyophilized powder
- Storage buffer:0,01M PBS, pH 7,2.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:500 µg
- Pk:500 µG
Specifications
About this item
Type: Primary
Antigen: Mucin 16
Clonality: monoclonal
Clone: 9H545
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human