Anti-Mucin 1 Mouse Monoclonal Antibody [clone: 5F168]
Catalog # USBIM4700-05A
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Mucin 1
- Antigen symbol:MUC1
- Clonality:Monoclonal
- Clone:5F168
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,4, 1% BSA, 0,1% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified
- Size:1 ml
- Pk:1 mL
Specifications
About this item
Recommended dilution: Immunochemistry: 1:10
Type: Primary
Antigen: Mucin 1
Clonality: monoclonal
Clone: 5F168
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human