Anti-ABCC3 Mouse Monoclonal Antibody [clone: M3II-21]
Catalog # USBIM4688-23A
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:ATP Binding Cassette, sub-family C, member 3
- Antigen symbol:ABCC3
- Clonality:Monoclonal
- Clone:M3II-21
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b
- Western blot:Yes
- Format:Supernatant
- Form:liquid
- Amino acid number:830 to 949
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:1 ml
- Pk:1 mL
Specifications
About this item
Recommended dilution: Immunochemistry: 1:20-1:50; Western Blot: 1:20-1:50
Type: Primary
Antigen: ABCC3 (ATP-binding cassette, sub-family C (CFTR/MRP), member 3)
Clonality: monoclonal
Clone: M3II-21
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: