Anti-SYCE1L Mouse Monoclonal Antibody [clone: M2II-12]
USBIM4688-20C
- Antibody type:Primary
- Antigen name:synaptonemal complex central element Protein 1-like
- Antigen symbol:SYCE1L
- Clonality:Monoclonal
- Clone:M2II-12
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a
- Western blot:Yes
- Format:Supernatant
- Form:liquid
- Amino acid number:860 to 950
- Storage buffer:0,7%BSA, 0,1% sodium azide.
- Storage temperature:Cool
- Concentration:~250 ug Ig/ml,
- Shipping temperature:Ice
- Size:1 ml
- Pk:1 mL
Recommended dilution: Immunochemistry: 1:20-50;; Western Blot: 1:20-50
Type: Primary
Antigen: SYCE1L (synaptonemal complex central element protein 1-like)
Clonality: monoclonal
Clone: M2II-12
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: