Anti-MHC Class 2, DR Monomorphic Mouse Monoclonal Antibody [clone: 10F112]
Catalog # USBIM3887-46E
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:MHC Class 2, DR Monomorphic
- Clonality:Monoclonal
- Clone:10F112
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Reactivity:Sheep,Goat,Human,Bovine
- Form:liquid
- Storage buffer:PBS, pH 7,4, 0,09% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:250 µg
- Pk:250 µG
Specifications
About this item
Recommended dilution: Flow Cytometry: 1:50-1:200;
Type: Primary
Antigen: MHC Class 2, DR Monomorphic
Clonality: monoclonal
Clone: 10F112
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Bovine, Goat, Human, Sheep