Order Entry
Czech Republic
ContactUsLinkComponent
Anti-MHC Class 1, H2, KK Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 36-7-5]
Anti-MHC Class 1, H2, KK Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 36-7-5]
  USBIM3885-85G
Anti-MHC Class 1, H2, KK Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 36-7-5]
  USBIM3885-85G

 

  • Antibody type:
    Primary
  • Antigen name:
    MHC Class 1, H2, KK
  • Clonality:
    Monoclonal
  • Clone:
    36-7-5
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Mouse
  • Form:
    liquid
  • Storage buffer:
    PBS, 0,1% sodium azide
  • Storage temperature:
    Cool
  • Concentration:
    ~0,5 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified
  • Size:
    500 µg
  • Pk:
    500 µG

 

 

Type: Primary
Antigen: MHC Class 1, H2, KK
Clonality: monoclonal
Clone: 36-7-5
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Mouse