Zadání objednávky
Czech Republic
ContactUsLinkComponent
Anti-Matrix Metalloproteinase 14 Rabbit Polyclonal Antibody
Anti-Matrix Metalloproteinase 14 Rabbit Polyclonal Antibody
Catalog # USBIM2428-99C
Supplier:  US Biological
Anti-Matrix Metalloproteinase 14 Rabbit Polyclonal Antibody
Catalog # USBIM2428-99C
Supplier:  US Biological

Specifications

  • Druh protilátky:
    Primární
  • Název antigenu:
    Matrix Metalloproteinase 14
  • Symbol antigenu:
    MMP14
  • Klonovalita:
    Polyclonal
  • Spojení:
    Unconjugated
  • Hostitel:
    Rabbit
  • Izotyp:
    IgG
  • Reaktivita:
    Human,
    Rat
  • Western blot:
    Yes
  • Formulář:
    lyophilized powder
  • Počet aminokyselin:
    561 to 577
  • Skladovací pufr:
    PBS, 5% BSA, 0,05% thimerosal, 0,05% sodium azide.
  • Skladovací teplota:
    Cool
  • Koncentrace:
    ~1 mg/ml (after reconstitution)
  • Transportní teplota:
    Ice
  • Čištění:
    purified by immunoaffinity chromatography
  • Rozměry protilátky:
    100 µg
  • Bal.:
    100 µG

Specifications

About this item

Recommended dilution: Western Blot: 0.1-0.5 ?g/ml; Immunochemistry: 0.5-1 ?g/ml

Type: Primary
Antigen: Matrix Metalloproteinase 14
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat