Anti-Matrix Metalloproteinase 13 Mouse Monoclonal Antibody [clone: 0.N.452]
Catalog # USBIM2428-11A
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Matrix Metalloproteinase 13
- Antigen symbol:MMP13
- Clonality:Monoclonal
- Clone:0.N.452
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1 kappa
- Reactivity:Rat,Mouse
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,4. &also supplied with BSA and sodium azide. See M2428-11.
- Storage temperature:Cool
- Concentration:0,2 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:500 µl
- Pk:500 µl
Specifications
About this item
Recommended dilution: Western Blot: 1 ?g/ml; Immunoprecipitation: 2?g/mg
Type: Primary
Antigen: Matrix Metalloproteinase 13
Clonality: monoclonal
Clone: 0.N.452
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Mouse, Rat