Order Entry
Czech Republic
ContactUsLinkComponent
Anti-Matrix Metalloproteinase 12 Rabbit Polyclonal Antibody
Anti-Matrix Metalloproteinase 12 Rabbit Polyclonal Antibody
Catalog # USBIM2427-65A
Supplier:  US Biological
Anti-Matrix Metalloproteinase 12 Rabbit Polyclonal Antibody
Catalog # USBIM2427-65A
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Matrix Metalloproteinase 12
  • Antigen symbol:
    MMP12
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    PBS, 0,02% thimerosal, 30% glycerol.
  • Storage temperature:
    Cool
  • Concentration:
    ~0,5 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Recommended dilution: Western Blot: 0.5-4?g/ml

Type: Primary
Antigen: Matrix Metalloproteinase 12
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse