Anti-MAP2K6 Mouse Monoclonal Antibody [clone: 6A114]
USBIM2363-22C
- Antibody type:Primary
- Antigen name:Mitogen-activated protein kinase kinase 6
- Antigen symbol:MAP2K6
- Clonality:Monoclonal
- Clone:6A114
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,4, 0,1% sodium azide, before the addition of glycerol to 40%.
- Storage temperature:Cool
- Concentration:~0,25 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Recommended dilution: Western Blot: 1-3?g/ml; ELISA: 0.1-1 ?g/ml
Type: Primary
Antigen: MAP2K6 (mitogen-activated protein kinase kinase 6)
Clonality: monoclonal
Clone: 6A114
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human