Anti-MAP Kinase 13 Mouse Monoclonal Antibody [clone: 1E11]
Catalog # USBIM2362-51H
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Mitogen-activated protein kinase 13
- Antigen symbol:MAPK13
- Clonality:Monoclonal
- Clone:1E11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:251 to 365
- Storage buffer:PBS, pH 7,2.
- Storage temperature:Cool
- Concentration:~0,1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Recommended dilution: Immunofluorescence: 10 ?g/ml; Immunochemistry: 5 ?g/ml
Type: Primary
Antigen: MAP Kinase 13
Clonality: monoclonal
Clone: 100000000000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human