Order Entry
Czech Republic
ContactUsLinkComponent
Anti-MAP Kinase 13 Mouse Monoclonal Antibody [clone: 2D8]
Anti-MAP Kinase 13 Mouse Monoclonal Antibody [clone: 2D8]
Catalog # USBIM2362-51G
Supplier:  US Biological
undefined
Anti-MAP Kinase 13 Mouse Monoclonal Antibody [clone: 2D8]
Catalog # USBIM2362-51G
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Mitogen-activated protein kinase 13
  • Antigen symbol:
    MAPK13
  • Clonality:
    Monoclonal
  • Clone:
    2D8
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG2b kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Amino acid number:
    251 to 365
  • Storage buffer:
    PBS, pH 7,2.
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

Recommended dilution: Western Blot: 1:500-1:1000; Immunofluorescence: 10 ?g/ml; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: MAP Kinase 13
Clonality: monoclonal
Clone: 2D8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human