Anti-MAP Kinase 12 Rabbit Polyclonal Antibody (Biotin)
Catalog # USBIM2362-50C-BIOT
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Mitogen-activated protein kinase 12
- Antigen symbol:MAPK12
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:C-terminal
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Type: Primary
Antigen: MAP Kinase 12
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse