Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-MAP Kinase 10 Rabbit Monoclonal Antibody [clone: 10F662]
  USBIM2362-38C
Anti-MAP Kinase 10 Rabbit Monoclonal Antibody [clone: 10F662]
  USBIM2362-38C

 

  • Antibody type:
    Primary
  • Antigen name:
    MAP Kinase 10
  • Antigen symbol:
    MAPK10
  • Clonality:
    Monoclonal
  • Clone:
    10F662
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat
  • Western blot:
    Yes
  • Format:
    Supernatant
  • Form:
    liquid
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µl
  • Pk:
    100 µl

 

 

Recommended dilution: Western Blot: 1:500-1:2000

Type: Primary
Antigen: MAP Kinase 10
Clonality: monoclonal
Clone: 10F662
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat