Anti-MAP Kinase 10 Rabbit Polyclonal Antibody (Biotin)
USBIM2362-38B-BIOT
- Antibody type:Primary
- Antigen name:MAP Kinase 10
- Antigen symbol:MAPK10
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:N-terminal
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 µl
- Pk:200 µl
Type: Primary
Antigen: MAP Kinase 10
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse