Order Entry
Czech Republic
ContactUsLinkComponent
Anti-MAP Kinase p44/42 Rabbit Monoclonal Antibody [clone: 9B154]
Anti-MAP Kinase p44/42 Rabbit Monoclonal Antibody [clone: 9B154]
Catalog # USBIM2352-20E
Supplier:  US Biological
Anti-MAP Kinase p44/42 Rabbit Monoclonal Antibody [clone: 9B154]
Catalog # USBIM2352-20E
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    MAP Kinase p44/42
  • Clonality:
    Monoclonal
  • Clone:
    9B154
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Zebrafish,
    Drosophila,
    Pig,
    Monkey,
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Format:
    Supernatant
  • Form:
    liquid
  • Modification:
    phospho-Thr202, phospho-Tyr204
  • Storage buffer:
    10mMsodium HEPES, pH 7,5, 150mM sodium chloride, 0,1mg/ml BSA, 0,02% sodium azide, 50% glycerol.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    200 µl
  • Pk:
    200 µl

Specifications

About this item

Recommended dilution: Western Blot: 1:1000; Immunoprecipitation: 1:50; Immunochemistry: 1:100

Type: Primary
Antigen: MAP Kinase p44/42
Clonality: monoclonal
Clone: 9B154
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Drosophila, Human, Monkey, Mouse, Pig, Rat, Zebrafish