Order Entry
Czech Republic
ContactUsLinkComponent
Anti-MAP Kinase p44/42 Rabbit Monoclonal Antibody [clone: 10F648]
Anti-MAP Kinase p44/42 Rabbit Monoclonal Antibody [clone: 10F648]
Catalog # USBIM2352-10D
Supplier:  US Biological
Anti-MAP Kinase p44/42 Rabbit Monoclonal Antibody [clone: 10F648]
Catalog # USBIM2352-10D
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    MAP Kinase p44/42
  • Clonality:
    Monoclonal
  • Clone:
    10F648
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Format:
    Supernatant
  • Form:
    liquid
  • Modification:
    phospho-Thr185, phospho-Tyr187
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

Recommended dilution: Western Blot: 1:5000

Type: Primary
Antigen: MAP Kinase p44/42
Clonality: monoclonal
Clone: 10F648
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human