Anti-MAP Kinase 14 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Catalog # USBIM2352-03U-APC
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Mitogen-activated protein kinase 14
- Antigen symbol:MAPK14
- Clonality:Polyclonal
- Conjugation:APC (Allophycocyanin)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2, contains ~0,09% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Tested applications:Fluoresence ELISA
- Purification:purified by immunoaffinity chromatography
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Recommended dilution: FLISA: 1:1000; Western Blot: 1:50-1:100
Do not freeze!
Type: Primary
Antigen: MAP Kinase 14
Clonality: polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human