Order Entry
Czech Republic
ContactUsLinkComponent
Anti-MRC1 Mouse Monoclonal Antibody [clone: 15-2]
Anti-MRC1 Mouse Monoclonal Antibody [clone: 15-2]
Catalog # USBIM2256-02
Supplier:  US Biological
Anti-MRC1 Mouse Monoclonal Antibody [clone: 15-2]
Catalog # USBIM2256-02
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Mannose Receptor C Type 1
  • Antigen symbol:
    MRC1
  • Clonality:
    Monoclonal
  • Clone:
    15-2
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    PBS, 0,09% sodium azide.
  • Storage temperature:
    Cool
  • Concentration:
    ~0,1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified
  • Size:
    200 µg
  • Pk:
    200 µG

Specifications

About this item

Type: Primary
Antigen: MRC1 (mannose receptor, C type 1)
Clonality: monoclonal
Clone: 15-2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human