Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-KDM1A Rabbit Polyclonal Antibody
  USBIL4595-50Q
Anti-KDM1A Rabbit Polyclonal Antibody
  USBIL4595-50Q

 

  • Antibody type:
    Primary
  • Antigen name:
    lysine (K)-specific demethylase 1A
  • Antigen symbol:
    KDM1A
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Dog,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Amino acid number:
    800 to 850
  • Storage buffer:
    PBS, 0,2% gelatin, 0,05% sodium azide.
  • Storage temperature:
    Cool
  • Concentration:
    ~0,5 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein G chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Recommended dilution: Western Blot: 1-3?g/ml; Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: KDM1A (lysine (K)-specific demethylase 1A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Dog, Human, Mouse, Rat