Anti-Lck Interacting Molecule Mouse Monoclonal Antibody [clone: LIME-10]
Catalog # USBIL1566-09
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Lck Interacting Molecule
- Clonality:Monoclonal
- Clone:LIME-10
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,4, 15mM sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: Lck Interacting Molecule
Clonality: monoclonal
Clone: LIME-10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human