Anti-LIME1 Mouse Monoclonal Antibody [clone: LIME-10]
USBIL1566-02A
- Antibody type:Primary
- Antigen name:Lck interacting transmembrane adaptor 1
- Antigen symbol:LIME1
- Clonality:Monoclonal
- Clone:LIME-10
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:281 to 296
- Storage buffer:PBS, 15mM sodium azide, pH 7,4.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:50 µg
- Pk:50 µG
Recommended dilution: Immunochemistry: 10 ?g/ml
Type: Primary
Antigen: LIME1 (Lck interacting transmembrane adaptor 1)
Clonality: monoclonal
Clone: LIME-10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human