Anti-LCK Rabbit Polyclonal Antibody
USBIL1565-15D
- Antibody type:Primary
- Antigen name:lymphocyte-specific Protein tyrosine kinase
- Antigen symbol:LCK
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Modification:phospho-Tyr505
- Storage buffer:PBS, pH 7,2, 1% BSA, 0,02% sodium azide, 50% glycerol.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Size:100 µg
- Pk:100 µG
Recommended dilution: Western Blot: 1-2?g/ml; Immunoprecipitation: 10-20 ?g/ml; Immunochemistry: 20-25 ?g/ml
Type: Primary
Antigen: LCK (lymphocyte-specific protein tyrosine kinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat