Order Entry
Czech Republic
ContactUsLinkComponent
Anti-Neural Cell Adhesion Molecule L1 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 9H148]
Anti-Neural Cell Adhesion Molecule L1 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 9H148]
Catalog # USBIL0010-07
Supplier:  US Biological
Anti-Neural Cell Adhesion Molecule L1 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 9H148]
Catalog # USBIL0010-07
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Neural Cell Adhesion Molecule L1
  • Clonality:
    Monoclonal
  • Clone:
    9H148
  • Conjugation:
    PE (Phycoerythrin)
  • Flow cytometry:
    Yes
  • Host:
    Rat
  • Isotype:
    IgG2a
  • Reactivity:
    Mouse
  • Form:
    liquid
  • Storage buffer:
    saline, 0,5% BSA, 0,1% sodium azide.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein G chromatography
  • Size:
    100 Tests
  • Pk:
    100 Tests

Specifications

About this item

Do not freeze!

Type: Primary
Antigen: Neural Cell Adhesion Molecule L1
Clonality: monoclonal
Clone: 9H148
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse