Anti-KRT18 Mouse Monoclonal Antibody [clone: RGE 53]
USBIK0199-21F
- Antibody type:Primary
- Antigen name:keratin 18
- Antigen symbol:KRT18
- Clonality:Monoclonal
- Clone:RGE 53
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Western blot:Yes
- Format:Supernatant
- Form:liquid
- Storage buffer:0,1% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:1 ml
- Pk:1 mL
Recommended dilution: Immunochemistry: 1:5-1:10
Type: Primary
Antigen: KRT18 (keratin 18)
Clonality: monoclonal
Clone: RGE 53
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: