Order Entry
Czech Republic
ContactUsLinkComponent
Anti-KRT7 Mouse Monoclonal Antibody [clone: OVTL12/30]
Anti-KRT7 Mouse Monoclonal Antibody [clone: OVTL12/30]
Catalog # USBIK0199-07H
Supplier:  US Biological
undefined
Anti-KRT7 Mouse Monoclonal Antibody [clone: OVTL12/30]
Catalog # USBIK0199-07H
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    keratin 7
  • Antigen symbol:
    KRT7
  • Clonality:
    Monoclonal
  • Clone:
    OVTL12/30
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Form:
    liquid
  • Storage buffer:
    PBS, 0,1% BSA, 0,1% sodium azide.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein G chromatography
  • Size:
    50 µl
  • Pk:
    50 µl

Specifications

About this item

Recommended dilution: Immunochemistry: 1:100

Type: Primary
Antigen: KRT7 (keratin 7)
Clonality: monoclonal
Clone: OVTL12/30
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human