Anti-KRT7 Mouse Monoclonal Antibody [clone: OVTL12/30]
USBIK0199-07C
- Antibody type:Primary
- Antigen name:keratin 7
- Antigen symbol:KRT7
- Clonality:Monoclonal
- Clone:OVTL12/30
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, 0,1% BSA, 0,1% sodium azide.
- Storage temperature:Cool
- Concentration:~0,2 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:1 ml
- Pk:1 mL
Type: Primary
Antigen: KRT7 (keratin 7)
Clonality: monoclonal
Clone: OVTL12/30
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human