Anti-Jumonji Domain Containing 2A Sheep Polyclonal Antibody
Catalog # USBIJ7876-45F
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Jumonji domain containing 2A
- Antigen symbol:JMJD2A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Sheep
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:lyophilized powder
- Storage buffer:PBSwith Trehalose.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Western Blot: 1 ?g/ml
Type: Primary
Antigen: Jumonji Domain Containing 2A
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human