Anti-IRTA5 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 7H13]
Catalog # USBII8711-30
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:IRTA5
- Clonality:Monoclonal
- Clone:7H13
- Conjugation:APC (Allophycocyanin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Form:liquid
- Storage buffer:PBS.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified
- Size:100 Tests
- Pk:100 Tests
Specifications
About this item
Do not freeze!
Type: Primary
Antigen: IRTA5
Clonality: monoclonal
Clone: 7H13
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human