Anti-IPO 38 Proliferation Marker Mouse Monoclonal Antibody [clone: 5H11]
Katalogové číslo USBII8460-01
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:IPO 38 Proliferation Marker
- Clonality:Monoclonal
- Clone:5H11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgM kappa
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,01% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: ELISA: 1:1000; Immunochemistry:1:50 to 1:100; Western Blot: 1:50 to 1:100
Type: Primary
Antigen: IPO 38 Proliferation Marker
Clonality: monoclonal
Clone: 5H11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM Kappa
Reactivity: Human, Mouse, Rat